missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMPR / GMPR1 Protein

A DNA sequence encoding the Homo sapiens (Human) GMPR / GMPR1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7200-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097670

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006868.3
GMPR / GMPR1 Protein
Research Use Only
GMPR
Recombinant Protein
E. coli
MPRIDADLKL DFKDVLLRPK RSSLKSRAEV DLERTFTFRN SKQTYSGIPI IVANMDTVGT FEMAAVMSQH SMFTAIHKHY SLDDWKLFAT NHPECLQNVA VSSGSGQNDL EKMTSILEAV PQVKFICLDV ANGYSEHFVE FVKLVRAKFP EHTIMAGNVV TGEMVEELIL SGADIIKVGV GPGSVCTTRT KTGVGYPQLS AVIECADSAH GLKGHIISDG GCTCPGDVAK AFGAGADFVM LGGMFSGHTE CAGEVFERNG RKLKLFYGMS SDTAMNKHAG GVAEYRASEG KTVEVPYKGD VENTILDILG GLRSTCTYVG AAKLKELSRR ATFIRVTQQH NTVFS
Greater than 90% as determined by SDS-PAGE.
2766
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers