missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMPR / GMPR1 Protein
A DNA sequence encoding the Homo sapiens (Human) GMPR / GMPR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7200-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006868.3 | |
GMPR / GMPR1 Protein | |
Research Use Only | |
GMPR | |
Recombinant Protein | |
E. coli | |
MPRIDADLKL DFKDVLLRPK RSSLKSRAEV DLERTFTFRN SKQTYSGIPI IVANMDTVGT FEMAAVMSQH SMFTAIHKHY SLDDWKLFAT NHPECLQNVA VSSGSGQNDL EKMTSILEAV PQVKFICLDV ANGYSEHFVE FVKLVRAKFP EHTIMAGNVV TGEMVEELIL SGADIIKVGV GPGSVCTTRT KTGVGYPQLS AVIECADSAH GLKGHIISDG GCTCPGDVAK AFGAGADFVM LGGMFSGHTE CAGEVFERNG RKLKLFYGMS SDTAMNKHAG GVAEYRASEG KTVEVPYKGD VENTILDILG GLRSTCTYVG AAKLKELSRR ATFIRVTQQH NTVFS | |
Greater than 90% as determined by SDS-PAGE. |
2766 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |