missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glypican-6 Protein

A DNA sequence encoding the Homo sapiens (Human) Glypican-6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9160-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007751

Quantity Price
1 2155.00 EUR / 1mg
3 2011.05 EUR / 1mg
Estimated Shipment
07-09-2020
Add to basket


Specifications

Specifications

NP_005699.1
Glypican-6 Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
10082
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GPC6
Recombinant Protein
Yeast
DVKARSCGEV RQAYGAKGFS LADIPYQEIA GEHLRICPQE YTCCTTEMED KLSQQSKLEF ENLVEETSHF VRTTFVSRHK KFDEFFRELL ENAEKSLNDM FVRTYGMLYM QNSEVFQDLF TELKRYYTGG NVNLEEMLND FWARLLERMF QLINPQYHFS EDYLECVSKY TDQLKPFGDV PRKLKIQVTR AFIAARTFVQ GLTVGREVAN RVSKVSPTPG CIRALMKMLY CPYCRGLPTV RPCNNYCLNV MKGCLANQAD LDTEWNLFID AMLLVAERLE GPFNIESVMD PIDVKISEAI MNMQENSMQV SAKVFQGCGQ PKPAPALRSA RSAPENFNTR FRPYNPEERP TTAAGTSLDR LVTDIKEKLK LSKKVWSALP YTICKDESVT AGTSNEEECW NGHSKARYLP EIMNDGLTNQ INNPEVDVDI TRPDTFIRQQ IMALRVMTNK LKNAYNGNDV NFQDTSDESS GSGSGSGCMD DVCPTEFEFV TTEAPAVDPD RREVDS
Videos
Documents