missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glypican-6 Protein
A DNA sequence encoding the Homo sapiens (Human) Glypican-6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9160-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005699.1 | |
Glypican-6 Protein | |
Research Use Only | |
GPC6 | |
Recombinant Protein | |
Yeast | |
DVKARSCGEV RQAYGAKGFS LADIPYQEIA GEHLRICPQE YTCCTTEMED KLSQQSKLEF ENLVEETSHF VRTTFVSRHK KFDEFFRELL ENAEKSLNDM FVRTYGMLYM QNSEVFQDLF TELKRYYTGG NVNLEEMLND FWARLLERMF QLINPQYHFS EDYLECVSKY TDQLKPFGDV PRKLKIQVTR AFIAARTFVQ GLTVGREVAN RVSKVSPTPG CIRALMKMLY CPYCRGLPTV RPCNNYCLNV MKGCLANQAD LDTEWNLFID AMLLVAERLE GPFNIESVMD PIDVKISEAI MNMQENSMQV SAKVFQGCGQ PKPAPALRSA RSAPENFNTR FRPYNPEERP TTAAGTSLDR LVTDIKEKLK LSKKVWSALP YTICKDESVT AGTSNEEECW NGHSKARYLP EIMNDGLTNQ INNPEVDVDI TRPDTFIRQQ IMALRVMTNK LKNAYNGNDV NFQDTSDESS GSGSGSGCMD DVCPTEFEFV TTEAPAVDPD RREVDS | |
Greater than 90% as determined by SDS-PAGE. |
10082 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |