missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutathione S-transferase A2 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6123-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060980

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000837.3
Glutathione S-transferase A2 Protein
Research Use Only
GSTA2
Recombinant Protein
E. coli
MAEKPKLHYS NIRGRMESIR WLLAAAGVEF EEKFIKSAED LDKLRNDGYL MFQQVPMVEI DGMKLVQTRA ILNYIASKYN LYGKDIKEKA LIDMYIEGIA DLGEMILLLP FTQPEEQDAK LALIQEKTKN RYFPAFEKVL KSHGQDYLVG NKLSRADIHL VELLYYVEEL DSSLISSFPL LKALKTRISN LPTVKKFLQP GSPRKPPMDE KSLEESRKIF RF
Greater than 80% as determined by SDS-PAGE.
2939
50 μg
Not determined.
Full Length of BC002895
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers