missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6123-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000837.3 | |
Glutathione S-transferase A2 Protein | |
Research Use Only | |
GSTA2 | |
Recombinant Protein | |
E. coli | |
MAEKPKLHYS NIRGRMESIR WLLAAAGVEF EEKFIKSAED LDKLRNDGYL MFQQVPMVEI DGMKLVQTRA ILNYIASKYN LYGKDIKEKA LIDMYIEGIA DLGEMILLLP FTQPEEQDAK LALIQEKTKN RYFPAFEKVL KSHGQDYLVG NKLSRADIHL VELLYYVEEL DSSLISSFPL LKALKTRISN LPTVKKFLQP GSPRKPPMDE KSLEESRKIF RF | |
Greater than 80% as determined by SDS-PAGE. |
2939 | |
10 μg | |
Not determined. | |
Full Length of BC002895 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |