missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein
A DNA sequence encoding the Human GH1 / Growth hormone 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10147-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000506.2 | |
GH1 / Growth hormone 1 Protein | |
Research Use Only | |
GH1 | |
The ED50 as determined by a cell proliferation assay using rat Nb2-11 lymphoma cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
2688 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
FPTIPLSRLF DNAMLRAHRL HQLAFDTYQE FEEAYIPKEQ KYSFLQNPQT SLCFSESIPT PSNREETQQK SNLELLRISL LLIQSWLEPV QFLRSVFANS LVYGASDSNV YDLLKDLEEG IQTLMGRLED GSPRTGQIFK QTYSKFDTNS HNDDALLKNY GLLYCFRKDM DKVETFLRIV QCRSVEGSCG F | |
Greater than 98% as determined by SDS-PAGE and HPLC. |