missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Gem-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Gem-associated protein 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8052-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072381

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001007270.1
Gem-associated protein 1
Research Use Only
GEMIN7
Recombinant Protein
E. coli
MQTPVNIPVP VLRLPRGPDG FSRGFAPDGR RAPLRPEVPE IQECPIAQES LESQEQRARA ALRERYLRSL LAMVGHQVSF TLHEGVRVAA HFGATDLDVA NFYVSQLQTP IGVQAEALLR CSDIISYTFK P
Greater than 90% as determined by SDS-PAGE.
79760
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers