missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDAP2 Protein
A DNA sequence encoding the Homo sapiens (Human) GDAP2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8095-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129061.1 | |
GDAP2 Protein | |
Research Use Only | |
GDAP2 | |
Recombinant Protein | |
E. coli | |
MDPLGAPSQF VDVDTLPSWG DSCQDELNSS DTTAEIFQED TVRSPFLYNK DVNGKVVLWK GDVALLNCTA IVNTSNESLT DKNPVSESIF MLAGPDLKED LQKLKGCRTG EAKLTKGFNL AARFIIHTVG PKYKSRYRTA AESSLYSCYR NVLQLAKEQS MSSVGFCVIN SAKRGYPLED ATHIALRTVR RFLEIHGETI EKVVFAVSDL EEGTYQKLLP LYFPRSLKEE NRSLPYLPAD IGNAEGEPVV PERQIRISEK PGAPEDNQEE EDEGLGVDLS FIGSHAFARM EGDIDKQRKL ILQGQLSEAA LQKQHQRNYN RWLCQARSED LSDIASLKAL YQTGVDNCGR TVMVVVGRNI PVTLIDMDKA LLYFIHVMDH IAVKEYVLVY FHTLTSEYNH LDSDFLKKLY DVVDVKYKRN LKAVYFVHPT FRSKVSTWFF TTFSVSGLKD KIHHVDSLHQ LFSAISPEQI DFPPFVLEYD ARVRSTRSSP SPGMVY | |
Greater than 90% as determined by SDS-PAGE. |
54834 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |