missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-9 Protein

A DNA sequence encoding the Homo sapiens (Human) Galectin-9, was expressed in the hosts and tags indicated.

202.00 EUR - 2155.00 EUR

Specifications

Accession Number NP_001317092.1
Gene ID (Entrez) 3965
Name Galectin-9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16087901
View Documents Promotion Details
enQuireBio™
QP9244-YE-100UG
100 μg
565.00 EUR
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16027911
View Documents Promotion Details
enQuireBio™
QP9244-YE-500UG
500 μg
1525.00 EUR
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16097901
View Documents Promotion Details
enQuireBio™
QP9244-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16017911
View Documents Promotion Details
enQuireBio™
QP9244-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16037911
View Documents Promotion Details
enQuireBio™
QP9244-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16007911
View Documents Promotion Details
enQuireBio™
QP9244-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001317092.1
Galectin-9 Protein
Not determined.
Full Length of Isoform 2
Human
His
Tris-based buffer, 50% glycerol
3965
Research Use Only
LGALS9
Recombinant Protein
Yeast
MAFSGSQAPY LSPAVPFSGT IQGGLQDGLQ ITVNGTVLSS SGTRFAVNFQ TGFSGNDIAF HFNPRFEDGG YVVCNTRQNG SWGPEERKTH MPFQKGMPFD LCFLVQSSDF KVMVNGILFV QYFHRVPFHR VDTISVNGSV QLSYISFQNP RTVPVQPAFS TVPFSQPVCF PPRPRGRRQK PPGVWPANPA PITQTVIHTV QSAPGQMFST PAIPPMMYPH PAYPMPFITT ILGGLYPSKS ILLSGTVLPS AQRFHINLCS GNHIAFHLNP RFDENAVVRN TQIDNSWGSE ERSLPRKMPF VRGQSFSVWI LCEAHCLKVA VDG
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications