missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human G-CSF / CSF3 Protein

A DNA sequence encoding the Homo sapiens (Human) G-CSF / CSF3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8545-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000750.1
G-CSF / CSF3 Protein
Research Use Only
CSF3
Recombinant Protein
E. coli
VQEATPLGPA SSLPQSFLLK CLEQVRKIQG DGAALQEKLV SECATYKLCH PEELVLLGHS LGIPWAPLSS CPSQALQLAG CLSQLHSGLF LYQGLLQALE GISPELGPTL DTLQLDVADF ATTIWQQMEE LGMAPALQPT QGAMPAFASA FQRRAGGVLV ASHLQSFLEV SYRV
Greater than 90% as determined by SDS-PAGE.
1440
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers