missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fucose-1-phosphate guanylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Fucose-1-phosphate guanylyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9143-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186257.2 | |
Fucose-1-phosphate guanylyltransferase Protein | |
Research Use Only | |
FPGT | |
Recombinant Protein | |
Yeast | |
MAAARDPPEV SLREATQRKL RRFSELRGKL VARGEFWDIV AITAADEKQE LAYNQQLSEK LKRKELPLGV QYHVFVDPAG AKIGNGGSTL CALQCLEKLY GDKWNSFTIL LIHSGGYSQR LPNASALGKI FTALPLGNPI YQMLELKLAM YIDFPLNMNP GILVTCADDI ELYSIGEFEF IRFDKPGFTA LAHPSSLTIG TTHGVFVLDP FDDLKHRDLE YRSCHRFLHK PSIEKMYQFN AVCRPGNFCQ QDFAGGDIAD LKLDSDYVYT DSLFYMDHKS AKMLLAFYEK IGTLSCEIDA YGDFLQALGP GATVEYTRNT SNVIKEESEL VEMRQRIFHL LKGTSLNVVV LNNSKFYHIG TTEEYLFYFT SDNSLKSELG LQSITFSIFP DIPECSGKTS CIIQSILDSR CSVAPGSVVE YSRLGPDVSV GENCIISGSY ILTKAALPAH SFVCSLSLKM NRCLKYATMA FGVQDNLKKS VKTLSDIKLL QFFGVCFLSC LDVWNLKVTE ELFSGNKTCL SLWTARIFPV CSSLSDSVIT SLKMLNAVKN KSAFSLNSYK LLSIEEMLIY KDVEDMITYR EQIFLEISLK SSLM | |
Greater than 90% as determined by SDS-PAGE. |
8790 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |