missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FOXP3 / Scurfin Protein

A DNA sequence encoding the Homo sapiens (Human) FOXP3 / Scurfin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8828-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001107849.1
FOXP3 / Scurfin Protein
Research Use Only
FOXP3
Recombinant Protein
E. coli
MPNPRPGKPS APSLALGPSP GASPSWRAAP KASDLLGARG PGGTFQGRDL RGGAHASSSS LNPMPPSQLQ LPTLPLVMVA PSGARLGPLP HLQALLQDRP HFMHQLSTVD AHARTPVLQV HPLESPAMIS LTPPTTATGV FSLKARPGLP PGINVASLEW VSREPALLCT FPNPSAPRKD STLSAVPQSS YPLLANGVCK WPGCEKVFEE PEDFLKHCQA DHLLDEKGRA QCLLQREMVQ SLEQQLVLEK EKLSAMQAHL
Greater than 90% as determined by SDS-PAGE.
50943
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers