missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroblast growth factor 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Fibroblast growth factor 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8665-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004455.2 | |
Fibroblast growth factor 5 Protein | |
Research Use Only | |
FGF5 | |
Recombinant Protein | |
E. coli | |
AWAHGEKRLA PKGQPGPAAT DRNPRGSSSR QSSSSAMSSS SASSSPAASL GSQGSGLEQS SFQWSPSGRR TGSLYCRVGI GFHLQIYPDG KVNGSHEANM LSVLEIFAVS QGIVGIRGVF SNKFLAMSKK GKLHASAKFT DDCKFRERFQ ENSYNTYASA IHRTEKTGRE WYVALNKRGK AKRGCSPRVK PQHISTHFLP RFKQSEQPEL SFTVTVPEKK KPPSPIKPKI PLSAPRKNTN SVKYRLKFRF G | |
Greater than 90% as determined by SDS-PAGE. |
2250 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |