missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Exportin-2 Protein
A DNA sequence encoding the Homo sapiens(Human) Exportin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9974-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243064.1 | |
Exportin-2 Protein | |
Research Use Only | |
CSE1L | |
Recombinant Protein | |
E. coli | |
MELSDANLQT LTEYLKKTLD PDPAIRRPAE KFLESVEGNQ NYPLLLLTLL EKSQDNVIKV CASVTFKNYI KRNWRIVEDE PNKICEADRV AIKANIVHLM LSSPEQIQKQ LSDAISIIGR EDFPQKWPDL LTEMVNRFQS GDFHVINGVL RTAHSLFKRY RHEFKSNELW TEIKLVLDAF ALPLTNLFKA TIELCSTHAN | |
Greater than 90% as determined by SDS-PAGE. |
1434 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |