missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Excitatory amino acid transporter 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6690-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_004161.4
Excitatory amino acid transporter 3 Protein
Research Use Only
SLC1A1
Recombinant Protein
E. coli
AEDVTLIIAV DWLLDRFRTM VNVLGDAFGT GIVEKLSKKE LEQMDVSSEV NIVNPFALES TILDNEDSDT KKSYVNGGFA VDKSDTISFT QTSQF
Greater than 90% as determined by SDS-PAGE.
6505
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers