missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Excitatory amino acid transporter 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6690-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004161.4 | |
Excitatory amino acid transporter 3 Protein | |
Research Use Only | |
SLC1A1 | |
Recombinant Protein | |
E. coli | |
AEDVTLIIAV DWLLDRFRTM VNVLGDAFGT GIVEKLSKKE LEQMDVSSEV NIVNPFALES TILDNEDSDT KKSYVNGGFA VDKSDTISFT QTSQF | |
Greater than 90% as determined by SDS-PAGE. |
6505 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |