missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Eukaryotic translation initiation factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Eukaryotic translation initiation factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8492-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074921

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005792.1
Eukaryotic translation initiation factor 1 Protein
Research Use Only
SUI1
Recombinant Protein
E. coli
MSAIQNLHSF DPFADASKGD DLLPAGTEDY IHIRIQQRNG RKTLTTVQGI ADDYDKKKLV KAFKKKFACN GTVIEHPEYG EVIQLQGDQR KNICQFLVEI GLAKDDQLKV HGF
Greater than 90% as determined by SDS-PAGE.
10209
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions