missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ES1 protein homolog, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) ES1 protein homolog, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8386-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044351

  • 2090.00 EUR / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004640.4
ES1 protein homolog, mitochondrial Protein
Research Use Only
C21orf33
Recombinant Protein
E. coli
ARVALVLSGC GVYDGTEIHE ASAILVHLSR GGAEVQIFAP DVPQMHVIDH TKGQPSEGES RNVLTESARI ARGKITDLAN LSAANHDAAI FPGGFGAAKN LSTFAVDGKD CKVNKEVERV LKEFHQAGKP IGLCCIAPVL AAKVLRGVEV TVGHEQEEGG KWPYAGTAEA IKALGAKHCV KEVVEAHVDQ KNKVVTTPAF MCETALHYIH DGIGAMVRKV LELTGK
Greater than 90% as determined by SDS-PAGE.
102724023
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers