missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ENPEP / Aminopeptidase A Protein

A DNA sequence encoding the Homo sapiens (Human) ENPEP / Aminopeptidase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8812-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006621

  • 2090.00 EUR / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001968.3
ENPEP / Aminopeptidase A Protein
Research Use Only
ENPEP
Recombinant Protein
E. coli
YFQGQVKPIA DSLGWNDAGD HVTKLLRSSV LGFACKMGDR EALNNASSLF EQWLNGTVSL PVNLRLLVYR YGMQNSGNEI SWNYTLEQYQ KTSLAQEKEK LLYGLASVKN VTLLSRYLDL LKDTNLIKTQ DVFTVIRYIS YNSYGKNMAW NWIQLNWDYL VNRYTLNNRN LGRIVTIAEP FNTELQLWQM ESFFAKYPQA GAGEKPREQV LETVKNNIEW LKQHRNTIRE W
Greater than 90% as determined by SDS-PAGE.
2028
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers