missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8402-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003312.3 | |
Elongation factor Tu, mitochondrial Protein | |
Research Use Only | |
TUFM | |
Recombinant Protein | |
E. coli | |
EAKKTYVRDK PHVNVGTIGH VDHGKTTLTA AITKILAEGG GAKFKKYEEI DNAPEERARG ITINAAHVEY STAARHYAHT DCPGHADYVK NMITGTAPLD GCILVVAAND GPMPQTREHL LLARQIGVEH VVVYVNKADA VQDSEMVELV ELEIRELLTE FGYKGEETPV IVGSALCALE GRDPELGLKS VQKLLDAVDT YIPVPARDLE KPFLLPVEAV YSVPGRGTVV TGTLERGILK KGDEC | |
Greater than 90% as determined by SDS-PAGE. |
7284 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |