missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_003312.3 |
---|---|
Gene ID (Entrez) | 7284 |
Name | Elongation factor Tu, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006320
|
enQuireBio™
QP6855-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026320
|
enQuireBio™
QP6855-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016320
|
enQuireBio™
QP6855-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036320
|
enQuireBio™
QP6855-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046320
|
enQuireBio™
QP6855-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056320
|
enQuireBio™
QP6855-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003312.3 | |
Elongation factor Tu, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7284 | |
Research Use Only | |
TUFM | |
Recombinant Protein | |
E. coli | |
AVEAKKTYVR DKPHVNVGTI GHVDHGKTTL TAAITKILAE GGGAKFKKYE EIDNAPEERA RGITINAAHV EYSTAARHYA HTDCPGHADY VKNMITGTAP LDGCILVVAA NDGPMPQTRE HLLLARQIGV EHVVVYVNKA DAVQDSEMVE LVELEIRELL TEFGYKGEET PVIVGSALCA LEGRDPELGL KSVQKLLDAV DTYIPVPARD LEKPFLLPVE AVYSVPGRGT VVTGTLERGI LKKGDECELL GHSKNIRTVV TGIEMFHKSL ERAEAGDNLG ALVRGLKRED LRRGLVMVKP GSIKPHQKVE AQVYILSKEE GGRHKPFVSH FMPVMFSLTW DMACRIILPP EKELAMPGED LKFNLILRQP MILEKGQRFT LRDGNRTIGT GLVTNTLAMT EEEKNIKWG | |
Greater than 90% as determined by SDS-PAGE. |