missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELAV-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7563-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001411.2 | |
ELAV-like protein 1 | |
Research Use Only | |
ELAVL3 | |
Recombinant Protein | |
Yeast | |
MVTQILGAME SQVGGGPAGP ALPNGPLLGT NGATDDSKTN LIVNYLPQNM TQDEFKSLFG SIGDIESCKL VRDKITGQSL GYGFVNYSDP NDADKAINTL NGLKLQTKTI KVSYARPSSA SIRDANLYVS GLPKTMSQKE MEQLFSQYGR IITSRILVDQ VTGVSRGVGF IRFDKRIEAE EAIKGLNGQK PLGAAEPITV KFANNPSQKT GQALLTHLYQ SSARRYAGPL HHQTQRFRLD NLLNMAYGVK SPLSLIARFS PIAIDGMSGL AGVGLSGGAA GAGWCIFVYN LSPEADESVL WQLFGPFGAV TNVKVIRDFT TNKCKGFGFV TMTNYDEAAM AIASLNGYRL GERVLQVSFK TSKQHKA | |
Greater than 90% as determined by SDS-PAGE. |
1995 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |