missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ELAV-like protein 1

A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7563-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029420

  • 1525.00 EUR / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001411.2
ELAV-like protein 1
Research Use Only
ELAVL3
Recombinant Protein
Yeast
MVTQILGAME SQVGGGPAGP ALPNGPLLGT NGATDDSKTN LIVNYLPQNM TQDEFKSLFG SIGDIESCKL VRDKITGQSL GYGFVNYSDP NDADKAINTL NGLKLQTKTI KVSYARPSSA SIRDANLYVS GLPKTMSQKE MEQLFSQYGR IITSRILVDQ VTGVSRGVGF IRFDKRIEAE EAIKGLNGQK PLGAAEPITV KFANNPSQKT GQALLTHLYQ SSARRYAGPL HHQTQRFRLD NLLNMAYGVK SPLSLIARFS PIAIDGMSGL AGVGLSGGAA GAGWCIFVYN LSPEADESVL WQLFGPFGAV TNVKVIRDFT TNKCKGFGFV TMTNYDEAAM AIASLNGYRL GERVLQVSFK TSKQHKA
Greater than 90% as determined by SDS-PAGE.
1995
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers