missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EFCAB14 Protein
A DNA sequence encoding the Homo sapiens (Human) EFCAB14, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_055589.1 |
---|---|
Gene ID (Entrez) | 9813 |
Name | EFCAB14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024831
|
enQuireBio™
QP8477-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044831
|
enQuireBio™
QP8477-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034831
|
enQuireBio™
QP8477-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054831
|
enQuireBio™
QP8477-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064831
|
enQuireBio™
QP8477-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074831
|
enQuireBio™
QP8477-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055589.1 | |
EFCAB14 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9813 | |
Research Use Only | |
EFCAB14 | |
Recombinant Protein | |
E. coli | |
MKKRKELNAL IGLAGDSRRK KPKKGPSSHR LLRTEPPDSD SESSSEEEEE FGVVGNRSRF AKGDYLRCCK ICYPLCGFVI LAACVVACVG LVWMQVALKE DLDALKEKFR TMESNQKSSF QEIPKLNEEL LSKQKQLEKI ESGEMGLNKV WINITEMNKQ ISLLTSAVNH LKANVKSAAD LISLPTTVEG LQKSVASIGN TLNSVHLAVE ALQKTVDEHK KTMELLQSDM NQHFLKETPG SNQIIPSPSA TSELDNKTHS ENLKQDILYL HNSLEEVNSA LVGYQRQNDL KLEGMNETVS NLTQRVNLIE SDVVAMSKVE KKANLSFSMM GDRSATLKRQ SLDQVTNRTD TVKIQSIKKE DSSNSQVSKL REKLQLISAL TNKPESNRPP ETADEEQVES FTSKPSALPK FSQFLGDPVE KAAQLRPISL PGVSSTEDLQ DLFRKTGQDV DGKLTYQEIW TSLGSAMPEP ESLRAFDSDG DGR | |
Greater than 90% as determined by SDS-PAGE. |