missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EF1B / EEF1B2 Protein
A DNA sequence encoding the Homo sapiens (Human) EF1B / EEF1B2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7171-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001032752.1 | |
EF1B / EEF1B2 Protein | |
Research Use Only | |
EEF1B2 | |
Recombinant Protein | |
E. coli | |
GFGDLKSPAG LQVLNDYLAD KSYIEGYVPS QADVAVFEAV SSPPPADLCH ALRWYNHIKS YEKEKASLPG VKKALGKYGP ADVEDTTGSG ATDSKDDDDI DLFGSDDEEE SEEAKRLREE RLAQYESKKA KKPALVAKSS ILLDVKPWDD ETDMAKLEEC VRSIQADGLV WGSSKLVPVG YGIKKLQIQC VVEDDKVGTD MLEEQITAFE DYVQSMDVAA FNKI | |
Greater than 80% as determined by SDS-PAGE. |
1933 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |