missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RNF114 Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RNF114, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6611-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024530

  • 525.00 EUR / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_061153.1
E3 ubiquitin-protein ligase RNF114 Protein
Research Use Only
RNF114
Recombinant Protein
E. coli
MAAQQRDCGG AAQLAGPAAE ADPLGRFTCP VCLEVYEKPV QVPCGHVFCS ACLQECLKPK KPVCGVCRSA LAPGVRAVEL ERQIESTETS CHGCRKNFFL SKIRSHVATC SKYQNYIMEG VKATIKDASL QPRNVPNRYT FPCPYCPEKN FDQEGLVEHC KLFHSTDTKS VVCPICASMP WGDPNYRSAN FREHIQRRHR FSYDTFVDYD VDEEDMMNQV LQRSIIDQ
Greater than 80% as determined by SDS-PAGE.
55905
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers