missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RNF114 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RNF114, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6611-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_061153.1 | |
E3 ubiquitin-protein ligase RNF114 Protein | |
Research Use Only | |
RNF114 | |
Recombinant Protein | |
E. coli | |
MAAQQRDCGG AAQLAGPAAE ADPLGRFTCP VCLEVYEKPV QVPCGHVFCS ACLQECLKPK KPVCGVCRSA LAPGVRAVEL ERQIESTETS CHGCRKNFFL SKIRSHVATC SKYQNYIMEG VKATIKDASL QPRNVPNRYT FPCPYCPEKN FDQEGLVEHC KLFHSTDTKS VVCPICASMP WGDPNYRSAN FREHIQRRHR FSYDTFVDYD VDEEDMMNQV LQRSIIDQ | |
Greater than 80% as determined by SDS-PAGE. |
55905 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |