missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA mismatch repair protein Msh6 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA mismatch repair protein Msh6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6388-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022950

  • 458.00 EUR / 100µg
Estimated Shipment: 27-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000170.1
DNA mismatch repair protein Msh6 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2956
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MSH6
Recombinant Protein
E. coli
MSRQSTLYSF FPKSPALSDA NKASARASRE GGRAAAAPGA SPSPGGDAAW SEAGPGPRPL ARSASPPKAK NLNGGLRRSV APAAPTSCDF SPGDLVWAKM EGYPWWPCLV YNHPFDGTFI REKGKSVRVH VQFFDDSPTR GWVSKRLLKP YTGSKSKEAQ KGGHFYSAKP EILRAMQRAD EALNKDKIKR LELAVCDEPS EPEEEEEMEV GTTYVTDKSE EDNEIESEEE VQPKTQGSRR SSRQIKKRRV ISDSESDIGG SDVEFKPDTK EEGSSDEISS GVGDSESEGL NSPVKVARKR KRMVTGNGSL KRKSSRKETP SATKQATSIS SETKNTLRAF SAPQNSESQA HVSGGGDDSS RPTVWYHETL EWLKEEKRRD EHRRRPDHPD FDASTLYVPE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers