missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytosolic beta-glucosidase Protein
A DNA sequence encoding the Homo sapiens (Human) Cytosolic beta-glucosidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8196-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001121904.1 | |
Cytosolic beta-glucosidase Protein | |
Research Use Only | |
GBA3 | |
Recombinant Protein | |
E. coli | |
MAFPAGFGWA AATAAYQVEG GWDADGKGPC VWDTFTHQGG ERVFKNQTGD VACGSYTLWE EDLKCIKQLG LTHYRFSLSW SRLLPDGTTG FINQKAIQLD KVNLQVYCAW SLLDNFEWNQ GYSSRFGLFH VDFEDPARPR VPYTSAKEYA KIIRNNGLEA HL | |
Greater than 80% as determined by SDS-PAGE. |
57733 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |