missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 9 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8232-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001003684.1
Cytochrome b-c1 complex subunit 9 Protein
Research Use Only
UQCR10
Recombinant Protein
E. coli
AAATLTSKLY SLLFRRTSTF ALTIIVGVMF FERAFDQGAD AIYDHINEGK LWKHIKHKYE NK
Greater than 80% as determined by SDS-PAGE.
29796
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali