missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 9 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8232-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003684.1 | |
Cytochrome b-c1 complex subunit 9 Protein | |
Research Use Only | |
UQCR10 | |
Recombinant Protein | |
E. coli | |
AAATLTSKLY SLLFRRTSTF ALTIIVGVMF FERAFDQGAD AIYDHINEGK LWKHIKHKYE NK | |
Greater than 80% as determined by SDS-PAGE. |
29796 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |