missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Complement C3 Protein

A DNA sequence encoding the Homo sapiens (Human) Complement C3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8695-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006001

  • 463.00 EUR / 100µg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000055.2
Complement C3 Protein
Research Use Only
C3
Recombinant Protein
E. coli
YSIITPNILR LESEETMVLE AHDAQGDVPV TVTVHDFPGK KLVLSSEKTV LTPATNHMGN VTFTIPANRE FKSEKGRNKF VTVQATFGTQ VVEKVVLVSL QSGYLFIQTD KTIYTPGSTV LYRIFTVNHK LLPVGRTVMV NIENPEGIPV KQDSLSSQNQ LGVLPLSWDI PELVNMGQWK IRAYYENSPQ QVFSTEFEVK
Greater than 90% as determined by SDS-PAGE.
718
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers