missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMM domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) COMM domain-containing protein 4, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001271306.1 |
---|---|
Gene ID (Entrez) | 54939 |
Name | COMM domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024891
|
enQuireBio™
QP8487-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044891
|
enQuireBio™
QP8487-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034891
|
enQuireBio™
QP8487-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054891
|
enQuireBio™
QP8487-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064891
|
enQuireBio™
QP8487-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074891
|
enQuireBio™
QP8487-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001271306.1 | |
COMM domain-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
54939 | |
Research Use Only | |
COMMD4 | |
Recombinant Protein | |
E. coli | |
MRFRFCGDLD CPDWVLAEIS TLAKMSSVKL RLLCSQVLKE LLGQGIDYEK ILKLTADAKF ESGDVKATVA VLSFILSSAA KHSVDGESLS SELQQLGLPK EHAASLCRCY EEKQSPLQKH LRVCSLRMNR LAGVGWRVDY TLSSSLLQSV EEPMVHLRLE VAAAPGTPAQ PVAMSLSADK FQVLLAELKQ AQTLM | |
Greater than 90% as determined by SDS-PAGE. |