missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-2(IV) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-2(IV) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8734-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001837.2 | |
Collagen alpha-2(IV) chain Protein | |
Research Use Only | |
COL4A2 | |
Recombinant Protein | |
E. coli | |
VKHSQTDQEP MCPVGMNKLW SGYSLLYFEG QEKAHNQDLG LAGSCLARFS TMPFLYCNPG DVCYYASRND KSYWLSTTAP LPMMPVAEDE IKPYISRCSV CEAPAIAIAV HSQDVSIPHC PAGWRSLWIG YSFLMHTAAG DEGGGQSLVS PGSCLEDFRA TPFIECNGGR GTCHYYANKY SFWLTTIPEQ SFQGSPSADT LKAGLIRTHI SRCQVCMKNL | |
Greater than 90% as determined by SDS-PAGE. |
1284 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |