missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Collagen alpha-1(XI) chain Protein

A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XI) chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9074-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007581

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001177638.1
Collagen alpha-1(XI) chain Protein
Research Use Only
XI
Recombinant Protein
Yeast
GPMGLTGRPG PVGGPGSSGA KGESGDPGPQ GPRGVQGPPG PTGKPGKRGR PGADGGRGMP GEPGAKGDRG FDGLPGLPGD KGHRGERGPQ GPPGPPGDDG MRGEDGEIGP RGLPGEAGPR GLLGPRGTPG APGQPGMAGV DGPPGPKGNM GPQGEPGPPG QQGNPGPQ
Greater than 90% as determined by SDS-PAGE.
1301
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers