missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMT1A Protein
A DNA sequence encoding the Homo sapiens (Human) CKMT1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8676-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001015001.1 | |
CKMT1A Protein | |
Research Use Only | |
CKMT1A | |
Recombinant Protein | |
E. coli | |
ASERRRLYPP SAEYPDLRKH NNCMASHLTP AVYARLCDKT TPTGWTLDQC IQTGVDNPGH PFIKTVGMVA GDEETYEVFA DLFDPVIQER HNGYDPRTMK HTTDLDASKI RSGYFDERYV LSSRVRTGRS IRGLSLPPAC TRAERREVER VVVDALSGLK GDLAGRYYRL SEMTEAEQQQ LIDDHFLFDK PVSPLLTAAG MARDWPDARG IWHNNEKSFL IWVNEEDHTR VISMEKGGNM KRVFERFCRG LKEVERLIQE RGWEFMWNER LGYILTCPSN LGTGLRAGVH IKLPLLSKDS RFPKILENLR LQKRGTGGVD TAATGGVFDI SNLDRLGKSE VELVQLVIDG VNYLIDCERR LERGQDIRIP TPVIHTKH | |
Greater than 90% as determined by SDS-PAGE. |
1159 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |