missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKMT1A Protein

A DNA sequence encoding the Homo sapiens (Human) CKMT1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8676-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025911

  • 2095.00 EUR / 1mg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001015001.1
CKMT1A Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
1159
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CKMT1A
Recombinant Protein
E. coli
ASERRRLYPP SAEYPDLRKH NNCMASHLTP AVYARLCDKT TPTGWTLDQC IQTGVDNPGH PFIKTVGMVA GDEETYEVFA DLFDPVIQER HNGYDPRTMK HTTDLDASKI RSGYFDERYV LSSRVRTGRS IRGLSLPPAC TRAERREVER VVVDALSGLK GDLAGRYYRL SEMTEAEQQQ LIDDHFLFDK PVSPLLTAAG MARDWPDARG IWHNNEKSFL IWVNEEDHTR VISMEKGGNM KRVFERFCRG LKEVERLIQE RGWEFMWNER LGYILTCPSN LGTGLRAGVH IKLPLLSKDS RFPKILENLR LQKRGTGGVD TAATGGVFDI SNLDRLGKSE VELVQLVIDG VNYLIDCERR LERGQDIRIP TPVIHTKH
Videos
SDS
Documents
Certificates
Special Offers

Special Offers