missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cell division cycle protein 23 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Cell division cycle protein 23 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8237-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004652.2 | |
Cell division cycle protein 23 homolog Protein | |
Research Use Only | |
CDC23 | |
Recombinant Protein | |
E. coli | |
AAVAPVLSIN SDFSDLREIK KQLLLIAGLT RERGLLHSSK WSAELAFSLP ALPLAELQPP PPITEEDAQD MDAYTLAKAY FDVKEYDRAA HFLHGCNSKK AYFLYMYSRY LSGEKKKDDE TVDSLGPLEK GQVKNEALRE LRVELSKKHQ ARELDGFGLY LYGVVLRKLD LVKEAIDVFV EATHVLPLHW GAWLELCNLI TDKEMLKFLS LPDTWMKEFF LAHIYTELQL IEEALQKYQN LIDVGFSKSS YIVSQIAVAY HNIRDIDKAL SIFNELRKQD PYRIENMDTF SNLLYVRSMK SELSYLAHNL CEIDKYRVET CCVIGNYYSL RSQHEKAALY FQRALKLNPR YLGAWTLMGH EYMEMKNTSA AIQAYRHAIE VNKRDYRAWY GLGQTYEILK MPFYCLYYYR RAHQLRPNDS RMLVALGECY EKLNQLVEAK KCYWRAYAVG DVEKMALVKL AKLHEQLTES EQAAQCYIKY IQDIYSCGEI VEHLEESTAF RYLAQYYFKC KLWDEASTCA QKCCAFNDTR EEGKALLRQI LQLRNQGETP TTEVPAPFFL PASLSANNTP TRRVSPLN | |
Greater than 90% as determined by SDS-PAGE. |
8697 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |