missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cell division cycle protein 23 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Cell division cycle protein 23 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8237-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013471

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_004652.2
Cell division cycle protein 23 homolog Protein
Research Use Only
CDC23
Recombinant Protein
E. coli
AAVAPVLSIN SDFSDLREIK KQLLLIAGLT RERGLLHSSK WSAELAFSLP ALPLAELQPP PPITEEDAQD MDAYTLAKAY FDVKEYDRAA HFLHGCNSKK AYFLYMYSRY LSGEKKKDDE TVDSLGPLEK GQVKNEALRE LRVELSKKHQ ARELDGFGLY LYGVVLRKLD LVKEAIDVFV EATHVLPLHW GAWLELCNLI TDKEMLKFLS LPDTWMKEFF LAHIYTELQL IEEALQKYQN LIDVGFSKSS YIVSQIAVAY HNIRDIDKAL SIFNELRKQD PYRIENMDTF SNLLYVRSMK SELSYLAHNL CEIDKYRVET CCVIGNYYSL RSQHEKAALY FQRALKLNPR YLGAWTLMGH EYMEMKNTSA AIQAYRHAIE VNKRDYRAWY GLGQTYEILK MPFYCLYYYR RAHQLRPNDS RMLVALGECY EKLNQLVEAK KCYWRAYAVG DVEKMALVKL AKLHEQLTES EQAAQCYIKY IQDIYSCGEI VEHLEESTAF RYLAQYYFKC KLWDEASTCA QKCCAFNDTR EEGKALLRQI LQLRNQGETP TTEVPAPFFL PASLSANNTP TRRVSPLN
Greater than 90% as determined by SDS-PAGE.
8697
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers