missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM4 Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM4, was expressed in the hosts and tags indicated.
202.00 EUR - 2425.00 EUR
Specifications
Accession Number | NP_001808.2 |
---|---|
Gene ID (Entrez) | 1089 |
Name | CEACAM4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907939
|
enQuireBio™
QP5812-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927939
|
enQuireBio™
QP5812-YE-1MG |
1 mg |
2425.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917939
|
enQuireBio™
QP5812-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937939
|
enQuireBio™
QP5812-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947939
|
enQuireBio™
QP5812-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957939
|
enQuireBio™
QP5812-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001808.2 | |
CEACAM4 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1089 | |
Research Use Only | |
CEACAM4 | |
Recombinant Protein | |
Yeast | |
FTIEALPSSA AEGKDVLLLA CNISETIQAY YWHKGKTAEG SPLIAGYITD IQANIPGAAY SGRETVYPNG SLLFQNITLE DAGSYTLRTI NASYDSDQAT GQLHVHQNNV PGLPVGAVAG | |
Greater than 90% as determined by SDS-PAGE. |