missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD52 / CDW52 Protein
A DNA sequence encoding the Homo sapiens (Human) CD52 / CDW52, was expressed in the hosts and tags indicated.
287.00 EUR - 3575.00 EUR
Specifications
Accession Number | NP_001794.2 |
---|---|
Gene ID (Entrez) | 1043 |
Name | CD52 / CDW52 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097531
|
enQuireBio™
QP9049-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017541
|
enQuireBio™
QP9049-YE-1MG |
1 mg |
3575.00 EUR
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007541
|
enQuireBio™
QP9049-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027541
|
enQuireBio™
QP9049-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037541
|
enQuireBio™
QP9049-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047541
|
enQuireBio™
QP9049-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001794.2 | |
CD52 / CDW52 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1043 | |
Research Use Only | |
CD52 | |
Recombinant Protein | |
Yeast | |
GQNDTSQTSS PS | |
Greater than 90% as determined by SDS-PAGE. |