missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD45 / PTPRC Protein
A DNA sequence encoding the Homo sapiens (Human) CD45 / PTPRC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9356-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002829.3 | |
CD45 / PTPRC Protein | |
Research Use Only | |
PTPRC | |
Recombinant Protein | |
Yeast | |
QSPTPSPTGL TTAKMPSVPL SSDPLPTHTT AFSPASTFER ENDFSETTTS LSPDNTSTQV SPDSLDNASA FNTTGVSSVQ TPHLPTHADS QTPSAGTDTQ TFSGSAANAK LNPTPGSNAI SDVPGERSTA STFPTDPVSP LTTTLSLAHH SSAALPARTS NTTITANTSD AYLNASETTT LSPSGSAVIS TTTIATTPSK PTCDEKYANI TVDYLYNKET KLFTAKLNVN ENVECGNNTC TNNEVHNLTE CKNASVSISH NSCTAPDKTL ILDVPPGVEK FQLHDCTQVE KADTTICLKW KNIETFTCDT QNITYRFQCG NMIFDNKEIK LENLEPEHEY KCDSEILYNN HKFTNASKII KTDFGSPGEP QIIFCRSEAA HQGVITWNPP QRSFHNFTLC YIKETEKDCL NLDKNLIKYD LQNLKPYTKY VLSLHAYIIA KVQRNGSAAM CHFTTKSAPP SQVWNMTVSM TSDNSMHVKC RPPRDRNGPH ERYHLEVEAG NTLVRNESHK NCDFRVKDLQ YSTDYTFKAY FHNGDYPGEP FILHHSTSYN SK | |
Greater than 90% as determined by SDS-PAGE. |
5788 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |