missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD40L / CD154 / TNFSF5 Protein
A DNA sequence encoding the Homo sapiens (Human) CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8528-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000065.1 | |
CD40L / CD154 / TNFSF5 Protein | |
Research Use Only | |
CD40LG | |
Recombinant Protein | |
E. coli | |
HRRLDKIEDE RNLHEDFVFM KTIQRCNTGE RSLSLLNCEE IKSQFEGFVK DIMLNKEETK KENSFEMQKG DQNPQIAAHV ISEASSKTTS VLQWAEKGYY TMSNNLVTLE NGKQLTVKRQ GLYYIYAQVT FCSNREASSQ APFIASLCLK SPGRFERILL RAANTHSSAK PCGQQSIHLG GVFELQPGAS VFVNVTDPSQ VSHGTGFTSF GL | |
Greater than 90% as determined by SDS-PAGE. |
959 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |