missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD40L / CD154 / TNFSF5 Protein

A DNA sequence encoding the Homo sapiens (Human) CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8528-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055111

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000065.1
CD40L / CD154 / TNFSF5 Protein
Research Use Only
CD40LG
Recombinant Protein
E. coli
HRRLDKIEDE RNLHEDFVFM KTIQRCNTGE RSLSLLNCEE IKSQFEGFVK DIMLNKEETK KENSFEMQKG DQNPQIAAHV ISEASSKTTS VLQWAEKGYY TMSNNLVTLE NGKQLTVKRQ GLYYIYAQVT FCSNREASSQ APFIASLCLK SPGRFERILL RAANTHSSAK PCGQQSIHLG GVFELQPGAS VFVNVTDPSQ VSHGTGFTSF GL
Greater than 90% as determined by SDS-PAGE.
959
10 μg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers