missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD24 / Ly-52 / CD24A Protein
A DNA sequence encoding the Homo sapiens (Human) CD24 / Ly-52 / CD24A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8690-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001278666.1 | |
CD24 / Ly-52 / CD24A Protein | |
Research Use Only | |
CD24 | |
Recombinant Protein | |
E. coli | |
SETTTGTSSN SSQSTSNTGL APNPTNATTK AAGGALQSTA SLFVVSLSLL HLYS | |
Greater than 90% as determined by SDS-PAGE. |
100133941 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |