missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD146 / MCAM Protein
A DNA sequence encoding the Homo sapiens (Human) CD146 / MCAM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8689-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006491.2 | |
CD146 / MCAM Protein | |
Research Use Only | |
MCAM | |
Recombinant Protein | |
E. coli | |
LSQSQGNLSH VDWFSVHKEK RTLIFRVRQG QGQSEPGEYE QRLSLQDRGA TLALTQVTPQ DERIFLCQGK RPRSQEYRIQ LRVYKAPEEP NIQVNPLGIP VNSKEPEEVA TCVGRNGYPI PQVIWYKNGR PLKEEKNRVH IQSSQTVESS GLYTLQSILK AQLVKEDKDA QFYCELNYRL PSGNHMKESR EVTVPVFYPT EKVWLEVEPV GMLKEGDRVE IRCLADGNPP PHFSISKQNP STREAEEETT NDNGVLVLEP ARKEHSGRYE CQGLDLDTMI SLLSEPQELL VNYVSDVRVS PAAPERQEGS SLTLTCEAES SQDLEFQWLR EETGQVLERG PVLQLHDLKR EAGGGYRCVA SVPSIPGLNR TQLVNVAIFG PPWMAFKERK VWVKENMVLN LSCEASGHPR PTISWNVNGT ASEQDQDPQR VLSTLNVLVT PELLETGVEC TASNDLGKNT SILFLELVNL TTLTPDSNTT TGLSTSTASP HTRANSTSTE RKLPEPESRG VVIVAVIVCI LVLAVLGAVL YFLYKKGKLP CRRSGKQEIT LPPSRKSELV VEVKSDKLPE EMGLLQGSSG DKRAPGDQGE KYIDLRH | |
Greater than 90% as determined by SDS-PAGE. |
4162 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |