missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD106 / VCAM1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD106 / VCAM1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001069.1 |
---|---|
Gene ID (Entrez) | 7412 |
Name | CD106 / VCAM1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096470
|
enQuireBio™
QP6877-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016480
|
enQuireBio™
QP6877-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006480
|
enQuireBio™
QP6877-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026480
|
enQuireBio™
QP6877-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036480
|
enQuireBio™
QP6877-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046480
|
enQuireBio™
QP6877-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001069.1 | |
CD106 / VCAM1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7412 | |
Research Use Only | |
VCAM1 | |
Recombinant Protein | |
E. coli | |
FKIETTPESR YLAQIGDSVS LTCSTTGCES PFFSWRTQID SPLNGKVTNE GTTSTLTMNP VSFGNEHSYL CTATCESRKL EKGIQVEIYS FPKDPEIHLS GPLEAGKPIT VKCSVADVYP FDRLEIDLLK GDHLMKSQEF LEDADRKSLE TKSLEVTFTP VIEDIGKVLV CRAKLHIDEM DSVPTVRQAV KELQVYISPK NTVISVNPST KLQEGGSVTM TCSSEGLPAP EIFWSKKLDN GNLQHLSGNA TLTLIAMRME DSGIYVCEGV NLIGKNRKEV ELIVQEKPFT VEISPGPRIA AQIGDSVMLT CSVMGCESPS FSWRTQIDSP LSGKVRSEGT NSTLTLSPVS FENEHSYLCT VTCGHKKLEK GIQVELYSFP RDPEIEMSGG LVNGSSVTVS CKVPSVYPLD RLEIELLKGE TILENIEFLE DTDMKSLENK SLEMTFIPTI EDTGKALVCQ AKLHIDDMEF EPKQRQSTQT LYVNVAPRDT TVLVSPSSIL EEGSSVNMTC LSQGFPAPKI LWSRQLPNGE LQPLSENATL TLISTKMEDS GVYLCEGINQ AGRSRKEVEL IIQVTPKDIK LTAFPSESVK EGDTVIISCT CGNVPETWII LKKKAETGDT VLKSIDGAYT IRKAQLKDAG VYECESKNKV GSQLRSLTLD VQGRENNKDY FSPE | |
Greater than 90% as determined by SDS-PAGE. |