missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD10 / Neprilysin / MME Protein
A DNA sequence encoding the Homo sapiens (Human) CD10 / Neprilysin / MME, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9276-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000893.2 | |
CD10 / Neprilysin / MME Protein | |
Research Use Only | |
MME | |
Recombinant Protein | |
Yeast | |
YDDGICKSSD CIKSAARLIQ NMDATTEPCT DFFKYACGGW LKRNVIPETS SRYGNFDILR DELEVVLKDV LQEPKTEDIV AVQKAKALYR SCINESAIDS RGGEPLLKLL PDIYGWPVAT ENWEQKYGAS WTAEKAIAQL NSKYGKKVLI NLFVGTDDKN SVNHVIHIDQ PRLGLPSRDY YECTGIYKEA CTAYVDFMIS VARLIRQEER LPIDENQLAL EMNKVMELEK EIANATAKPE DRNDPMLLYN KMTLAQIQNN FSLEINGKPF SWLNFTNEIM STVNISITNE EDVVVYAPEY LTKLKPILTK YSARDLQNLM SWRFIMDLVS SLSRTYKESR NAFRKALYGT TSETATWRRC ANYVNGNMEN AVGRLYVEAA FAGESKHVVE DLIAQIREVF IQTLDDLTWM DAETKKRAEE KALAIKERIG YPDDIVSNDN KLNNEYLELN YKEDEYFENI IQNLKFSQSK QLKKLREKVD KDEWISGAAV VNAFYSSGRN QIVFPAGILQ PPFFSAQQSN SLNYGGIGMV IGHEITHGFD DNGRNFNKDG DLVDWWTQQS ASNFKEQSQC MVYQYGNFSW DLAGGQHLNG INTLGENIAD NGGLGQAYRA YQNYIKKNGE EKLLPGLDLN HKQLFFLNFA QVWCGTYRPE YAVNSIKTDV HSPGNFRIIG TLQNSAEFSE AFHCRKNSYM NPEKKCRVW | |
Greater than 90% as determined by SDS-PAGE. |
4311 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |