missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL5 / RANTES Protein

A DNA sequence encoding the Homo sapiens (Human) CCL5 / RANTES, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8524-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025101

  • 458.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002976.2
CCL5 / RANTES Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6352
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CCL5
Recombinant Protein
E. coli
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers